Learn More
Meso Scale Discovery U-PLEX Mouse VEGF-a ASY
SDP
Catalog No. 5000078358
Encompass
The Meso Scale Discovery U-PLEX Mouse VEGF-A Assay Kit is a multiplex immunoassay for measuring mouse VEGF-A in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers It quantifies mouse VEGF-A concentrations from 0 53-2000 pg/mL with minimal sample volume aiding in biomarker research
Catalog No. 50-000-78358 Supplier Meso Scale Discovery Supplier No. K152UVK1
The Meso Scale Discovery U-PLEX Mouse VEGF-A Assay Kit is a multiplex immunoassay for measuring mouse VEGF-A in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers It quantifies mouse VEGF-A concentrations from 0 53-2000 pg/mL with minimal sample volume aiding in biomarker research