Learn More
Meso Scale Discovery U-PLEX NHP CTACK ASY
SDP
Catalog No. 5000079746
Encompass
The Meso Scale Discovery U-PLEX NHP CTACK Assay Kit is a multiplex immunoassay for measuring CTACK levels in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG reagents calibrators and controls It has a sensitivity of 0 08 pg/mL and an upper limit of 2500 pg/mL enabling high-throughput sensitive and accurate CTACK quantification in research
Catalog No. 50-000-79746 Supplier Meso Scale Discovery Supplier No. K156VDK2
The Meso Scale Discovery U-PLEX NHP CTACK Assay Kit is a multiplex immunoassay for measuring CTACK levels in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG reagents calibrators and controls It has a sensitivity of 0 08 pg/mL and an upper limit of 2500 pg/mL enabling high-throughput sensitive and accurate CTACK quantification in research