Learn More
Meso Scale Discovery U-PLEX NHP CTACK ASY
SDP
Catalog No. 5000080044
Encompass
The Meso Scale Discovery U-PLEX NHP CTACK Assay Kit is a multiplex immunoassay that measures CTACK (Cutaneous T-Cell Attracting Chemokine) levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and diluent This allows for the accurate quantitative measurement of CTACK levels with high sensitivity and a broad dynamic range The kit provides all necessary reagents to complete the assay
Catalog No. 50-000-80044 Supplier Meso Scale Discovery Supplier No. K156VDK1
The Meso Scale Discovery U-PLEX NHP CTACK Assay Kit is a multiplex immunoassay that measures CTACK (Cutaneous T-Cell Attracting Chemokine) levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and diluent This allows for the accurate quantitative measurement of CTACK levels with high sensitivity and a broad dynamic range The kit provides all necessary reagents to complete the assay