Learn More
Meso Scale Discovery U-PLEX NHP CTACK ASY384-well
SDP
Catalog No. 5000080223
Encompass
The Meso Scale Discovery U-PLEX NHP CTACK 384-Well Assay Kit is a 384-well multiplex immunoassay that measures cutaneous T-cell-attracting chemokine (CTACK) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls to generate a standard curve and validate assay performance It allows high-throughput quantitative determination of CTACK levels with minimal hands-on time
Catalog No. 50-000-80223 Supplier Meso Scale Discovery Supplier No. K256VDK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP CTACK 384-Well Assay Kit is a 384-well multiplex immunoassay that measures cutaneous T-cell-attracting chemokine (CTACK) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls to generate a standard curve and validate assay performance It allows high-throughput quantitative determination of CTACK levels with minimal hands-on time