Learn More
Meso Scale Discovery U-PLEX NHP CTACK ASYQP 25 PL
SDP
Catalog No. 5000080036
Encompass
The MSD U-PLEX NHP CTACK Assay (K156VDK) is a multiplex immunoassay kit designed for quantitative measurement of CTACK (CCL27) biomarker in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit includes pre-coated plates detection reagents and calibrators for reliable reproducible results in cytokine analysis studies
Catalog No. 50-000-80036 Supplier Meso Scale Discovery Supplier No. K156VDK24
Add to Cart
Add to Cart
Request a Quote
The MSD U-PLEX NHP CTACK Assay (K156VDK) is a multiplex immunoassay kit designed for quantitative measurement of CTACK (CCL27) biomarker in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit includes pre-coated plates detection reagents and calibrators for reliable reproducible results in cytokine analysis studies