Learn More
Meso Scale Discovery U-PLEX NHP ENA-78 ASY 25 PL
SDP
Catalog No. 5000079779
Encompass
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is an electrochemiluminescent immunoassay for measuring autoantibodies to 7 nuclear antigens (RNP Sm SS-A SS-B Scl-70 Jo-1 CENP-B) and 1 extractable nuclear antigen (dsDNA) in human serum plasma and other fluids This multiplex assay detects autoantibodies linked to systemic autoimmune diseases like lupus Sj gren s syndrome scleroderma polymyositis and mixed connective tissue disease using MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from small samples
Catalog No. 50-000-79779 Supplier Meso Scale Discovery Supplier No. K156VEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is an electrochemiluminescent immunoassay for measuring autoantibodies to 7 nuclear antigens (RNP Sm SS-A SS-B Scl-70 Jo-1 CENP-B) and 1 extractable nuclear antigen (dsDNA) in human serum plasma and other fluids This multiplex assay detects autoantibodies linked to systemic autoimmune diseases like lupus Sj gren s syndrome scleroderma polymyositis and mixed connective tissue disease using MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from small samples