Learn More
Meso Scale Discovery U-PLEX NHP ENA-78 ASY
SDP
Catalog No. 5000080033
Encompass
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is an electrochemiluminescent immunoassay for measuring autoantibodies to 7 nuclear antigens (RNP Sm SS-A SS-B Scl-70 Jo-1 CENP-B) and 1 extractable nuclear antigen (dsDNA) in human serum plasma and other fluids It detects autoantibodies linked to systemic autoimmune diseases like lupus Sj gren s syndrome scleroderma polymyositis and mixed connective tissue disease The assay uses MSD s QuickPlex SQ 120 and MULTI-ARRAY technology for rapid sensitive and specific results
Catalog No. 50-000-80033 Supplier Meso Scale Discovery Supplier No. K156VEK1
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is an electrochemiluminescent immunoassay for measuring autoantibodies to 7 nuclear antigens (RNP Sm SS-A SS-B Scl-70 Jo-1 CENP-B) and 1 extractable nuclear antigen (dsDNA) in human serum plasma and other fluids It detects autoantibodies linked to systemic autoimmune diseases like lupus Sj gren s syndrome scleroderma polymyositis and mixed connective tissue disease The assay uses MSD s QuickPlex SQ 120 and MULTI-ARRAY technology for rapid sensitive and specific results