Learn More
Meso Scale Discovery U-PLEX NHP ENA-78 ASY
SDP
Catalog No. 5000080071
Encompass
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is a multiplex immunoassay for detecting autoantibodies to 7 nuclear antigens and 1 extractable nuclear antigen from human samples It uses MSD s electrochemiluminescent technology offering a sensitive method for profiling with minimal sample volume The kit includes reagents and plates for 88 samples in duplicate plus calibrators and controls Results are in Units based on internal standards supporting efficient autoantibody detection for research and clinical use
Catalog No. 50-000-80071 Supplier Meso Scale Discovery Supplier No. K156VEK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP ENA 7-8 Assay Kit is a multiplex immunoassay for detecting autoantibodies to 7 nuclear antigens and 1 extractable nuclear antigen from human samples It uses MSD s electrochemiluminescent technology offering a sensitive method for profiling with minimal sample volume The kit includes reagents and plates for 88 samples in duplicate plus calibrators and controls Results are in Units based on internal standards supporting efficient autoantibody detection for research and clinical use