Learn More
Meso Scale Discovery U-PLEX NHP ENA-78 ASYQP
SDP
Catalog No. 5000080119
Encompass
The U-PLEX NHP ENA-78 Assay (K156VEK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of ENA-78 (CXCL5) protein levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring excellent reproducibility and wide dynamic range The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable biomarker analysis in research applications
Catalog No. 50-000-80119 Supplier Meso Scale Discovery Supplier No. K156VEK22
Add to Cart
Add to Cart
The U-PLEX NHP ENA-78 Assay (K156VEK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of ENA-78 (CXCL5) protein levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring excellent reproducibility and wide dynamic range The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable biomarker analysis in research applications