Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin-3 ASY
SDP
Catalog No. 5000078545
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in non-human primate serum plasma and other biological fluids This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results are obtained in as little as 3 5 hours using the MSD QuickPlex SQ 120 instrument This assay allows reliable quantification of Eotaxin-3 enabling research into its role in inflammatory processes and diseases
Catalog No. 50-000-78545 Supplier Meso Scale Discovery Supplier No. K156UEK1
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in non-human primate serum plasma and other biological fluids This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results are obtained in as little as 3 5 hours using the MSD QuickPlex SQ 120 instrument This assay allows reliable quantification of Eotaxin-3 enabling research into its role in inflammatory processes and diseases