Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin-3 ASY
SDP
Catalog No. 5000080825
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in non-human primate serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and Calibrator This assay allows for the quantitative measurement of Eotaxin-3 levels with high precision and reproducibility
Catalog No. 50-000-80825 Supplier Meso Scale Discovery Supplier No. K156UEK2
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in non-human primate serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and Calibrator This assay allows for the quantitative measurement of Eotaxin-3 levels with high precision and reproducibility