Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin ASY 25 PL
SDP
Catalog No. 5000081321
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay Kit is a multiplex immunoassay for measuring Eotaxin in non-human primate serum plasma and other fluids It uses electrochemiluminescence detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators controls 10X read and wash buffers and plate seals This assay quantifies Eotaxin aiding research into inflammatory and allergic responses in non-human primate models
Catalog No. 50-000-81321 Supplier Meso Scale Discovery Supplier No. K156UDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay Kit is a multiplex immunoassay for measuring Eotaxin in non-human primate serum plasma and other fluids It uses electrochemiluminescence detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators controls 10X read and wash buffers and plate seals This assay quantifies Eotaxin aiding research into inflammatory and allergic responses in non-human primate models