Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin ASY
SDP
Catalog No. 5000079905
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay is a multiplex immunoassay panel that measures Eotaxin levels in non-human primate serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has high sensitivity requires a small sample volume and provides results in under 3 hours This product can aid researchers studying immune response and inflammation in non-human primate models With minimal sample consumption rapid turnaround time and ultra-high sensitivity this Eotaxin assay panel enables efficient and robust analysis
Catalog No. 50-000-79905 Supplier Meso Scale Discovery Supplier No. K156UDK2
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay is a multiplex immunoassay panel that measures Eotaxin levels in non-human primate serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has high sensitivity requires a small sample volume and provides results in under 3 hours This product can aid researchers studying immune response and inflammation in non-human primate models With minimal sample consumption rapid turnaround time and ultra-high sensitivity this Eotaxin assay panel enables efficient and robust analysis