Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin ASY
SDP
Catalog No. 5000081320
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay Kit is a multiplex immunoassay for measuring Eotaxin in non-human primate samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls buffers and seals Sensitivity is 0 04 pg/mL with a quantification limit of 2500 pg/mL enabling accurate Eotaxin measurement for research
Catalog No. 50-000-81320 Supplier Meso Scale Discovery Supplier No. K156UDK1
The Meso Scale Discovery U-PLEX NHP Eotaxin Assay Kit is a multiplex immunoassay for measuring Eotaxin in non-human primate samples It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls buffers and seals Sensitivity is 0 04 pg/mL with a quantification limit of 2500 pg/mL enabling accurate Eotaxin measurement for research