Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin ASYQP
SDP
Catalog No. 5000080849
Encompass
The U-PLEX NHP Eotaxin Assay (K156UDK) is a multiplex immunoassay designed for quantitative measurement of eotaxin in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides reagents and protocols for detecting eotaxin levels in serum plasma and other biological matrices from non-human primate species making it valuable for preclinical studies and biomarker research
Catalog No. 50-000-80849 Supplier Meso Scale Discovery Supplier No. K156UDK21
The U-PLEX NHP Eotaxin Assay (K156UDK) is a multiplex immunoassay designed for quantitative measurement of eotaxin in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides reagents and protocols for detecting eotaxin levels in serum plasma and other biological matrices from non-human primate species making it valuable for preclinical studies and biomarker research