Learn More
Meso Scale Discovery U-PLEX NHP FLT3L ASY
SDP
Supplier:  Meso Scale Discovery K156XFK2
Encompass
The Meso Scale Discovery U-PLEX NHP FLT3L Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human FLT3 ligand in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results comparable to ELISA The assay has a detection range of 19 5-20 000 pg/mL and requires 50 L sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents for performing the assay It provides a high-throughput method to accurately quantify FLT3L levels
Catalog No. 50-000-78638