Learn More
Meso Scale Discovery U-PLEX NHP FLT3L ASY
SDP
Catalog No. 5000080100
Encompass
The Meso Scale Discovery U-PLEX NHP FLT3L Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human FLT3 ligand in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 1 5 pg/mL and the assay range is up to 10 000 pg/mL
Catalog No. 50-000-80100 Supplier Meso Scale Discovery Supplier No. K156XFK1
The Meso Scale Discovery U-PLEX NHP FLT3L Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human FLT3 ligand in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 1 5 pg/mL and the assay range is up to 10 000 pg/mL