Learn More
Meso Scale Discovery U-PLEX NHP FLT3L ASY384-well
SDP
Catalog No. 5000078457
Encompass
The Meso Scale Discovery U-PLEX NHP FLT3L 384-Well Assay Kit is a 384-well multiplex assay that measures human FLT3 ligand in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager and provides a testing capacity of up to 384 samples per plate It includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator The assay can detect FLT3 ligand concentrations from 0 08 pg/mL to 1000 pg/mL
Catalog No. 50-000-78457 Supplier Meso Scale Discovery Supplier No. K256XFK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP FLT3L 384-Well Assay Kit is a 384-well multiplex assay that measures human FLT3 ligand in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager and provides a testing capacity of up to 384 samples per plate It includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator The assay can detect FLT3 ligand concentrations from 0 08 pg/mL to 1000 pg/mL