Learn More
Meso Scale Discovery U-PLEX NHP Fractalkine ASY
SDP
Catalog No. 5000080047
Encompass
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit for measuring fractalkine in non-human primate samples like serum and plasma It uses MSD s electroluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation It detects fractalkine from 45 to 100 000 pg/mL enabling efficient analysis in NHP studies
Catalog No. 50-000-80047 Supplier Meso Scale Discovery Supplier No. K156VCK2
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit for measuring fractalkine in non-human primate samples like serum and plasma It uses MSD s electroluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation It detects fractalkine from 45 to 100 000 pg/mL enabling efficient analysis in NHP studies