Learn More
Meso Scale Discovery U-PLEX NHP Fractalkine ASY
SDP
Catalog No. 5000080082
Encompass
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of fractalkine in non-human primate (NHP) samples such as serum plasma and other biological fluids This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows for efficient reproducible analysis of fractalkine levels in NHP studies
Catalog No. 50-000-80082 Supplier Meso Scale Discovery Supplier No. K156VCK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP Fractalkine Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of fractalkine in non-human primate (NHP) samples such as serum plasma and other biological fluids This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows for efficient reproducible analysis of fractalkine levels in NHP studies