Learn More
Meso Scale Discovery U-PLEX NHP Fractalkine ASYQP
SDP
Catalog No. 5000080053
Encompass
The U-PLEX NHP Fractalkine Assay (K156VCK) is a specialized immunoassay designed for measuring fractalkine (CX3CL1) levels in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for quantitative detection of fractalkine a chemokine involved in immune cell recruitment and inflammation processes making it valuable for preclinical studies and biomarker research in primate models
Catalog No. 50-000-80053 Supplier Meso Scale Discovery Supplier No. K156VCK21
The U-PLEX NHP Fractalkine Assay (K156VCK) is a specialized immunoassay designed for measuring fractalkine (CX3CL1) levels in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for quantitative detection of fractalkine a chemokine involved in immune cell recruitment and inflammation processes making it valuable for preclinical studies and biomarker research in primate models