Learn More
Meso Scale Discovery U-PLEX NHP G-CSF ASY 25 PL
SDP
Catalog No. 5000080104
Encompass
The Meso Scale Discovery U-PLEX NHP CSF Assay is a multiplex immunoassay panel measuring neurofilament light chain total tau phosphorylated tau amyloid beta 42 amyloid beta 40 YKL-40 and neurogranin in human cerebrospinal fluid Utilizing electrochemiluminescence detection it provides quantitative results in under 3 hours with a small sample Intended for research use it aids in understanding neurodegenerative diseases like Alzheimer s allowing efficient measurement of key biomarkers from a single CSF sample
Catalog No. 50-000-80104 Supplier Meso Scale Discovery Supplier No. K156VGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP CSF Assay is a multiplex immunoassay panel measuring neurofilament light chain total tau phosphorylated tau amyloid beta 42 amyloid beta 40 YKL-40 and neurogranin in human cerebrospinal fluid Utilizing electrochemiluminescence detection it provides quantitative results in under 3 hours with a small sample Intended for research use it aids in understanding neurodegenerative diseases like Alzheimer s allowing efficient measurement of key biomarkers from a single CSF sample