Learn More
Meso Scale Discovery U-PLEX NHP G-CSF ASYQP 25 PL
SDP
Catalog No. 5000080997
Encompass
The U-PLEX NHP G-CSF Assay (K156VGK) is a multiplex immunoassay designed to measure Granulocyte Colony-Stimulating Factor (G-CSF) levels in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection The kit provides researchers with validated reagents and protocols for quantitative analysis of G-CSF in serum plasma and cell culture supernatants from primate species supporting preclinical research applications
Catalog No. 50-000-80997 Supplier Meso Scale Discovery Supplier No. K156VGK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP G-CSF Assay (K156VGK) is a multiplex immunoassay designed to measure Granulocyte Colony-Stimulating Factor (G-CSF) levels in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection The kit provides researchers with validated reagents and protocols for quantitative analysis of G-CSF in serum plasma and cell culture supernatants from primate species supporting preclinical research applications