Learn More
Meso Scale Discovery U-PLEX NHP GM-CSF ASY 25 PL
SDP
Catalog No. 5000080085
Encompass
The Meso Scale Discovery U-PLEX NHP GM-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human GM-CSF in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can accurately quantify GM-CSF levels between 0 06 - 1500 pg/mL with minimal cross-reactivity to related analytes The kit offers high performance immunoassays in a simple and rapid workflow
Catalog No. 50-000-80085 Supplier Meso Scale Discovery Supplier No. K156UMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP GM-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human GM-CSF in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can accurately quantify GM-CSF levels between 0 06 - 1500 pg/mL with minimal cross-reactivity to related analytes The kit offers high performance immunoassays in a simple and rapid workflow