Learn More
Meso Scale Discovery U-PLEX NHP GM-CSF ASY
SDP
Catalog No. 5000080051
Encompass
The Meso Scale Discovery U-PLEX NHP GM-CSF Assay is a multiplex immunoassay kit for measuring GM-CSF in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection technology on the QuickPlex SQ 120 instrument The assay detects 0 06-1500 pg/mL and shows minimal cross-reactivity with other cytokines The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents for testing enabling efficient high-sensitivity quantification of NHP GM-CSF
Catalog No. 50-000-80051 Supplier Meso Scale Discovery Supplier No. K156UMK2
The Meso Scale Discovery U-PLEX NHP GM-CSF Assay is a multiplex immunoassay kit for measuring GM-CSF in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection technology on the QuickPlex SQ 120 instrument The assay detects 0 06-1500 pg/mL and shows minimal cross-reactivity with other cytokines The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents for testing enabling efficient high-sensitivity quantification of NHP GM-CSF