Learn More
Meso Scale Discovery U-PLEX NHP GM-CSF ASY384-well
SDP
Catalog No. 5000081345
Encompass
The U-PLEX NHP GM-CSF 384-well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants It utilizes MSD s ultra-sensitive immunoassay technology and provides a lower limit of detection of 0 04 pg/mL with an upper limit of quantification of 1500 pg/mL The assay requires a low sample volume of 25 L and has a short 3 5 hour protocol It includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of lyophilized calibrator
Catalog No. 50-000-81345 Supplier Meso Scale Discovery Supplier No. K256UMK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP GM-CSF 384-well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants It utilizes MSD s ultra-sensitive immunoassay technology and provides a lower limit of detection of 0 04 pg/mL with an upper limit of quantification of 1500 pg/mL The assay requires a low sample volume of 25 L and has a short 3 5 hour protocol It includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of lyophilized calibrator