Learn More
Meso Scale Discovery U-PLEX NHP GM-CSF ASYQP
SDP
Catalog No. 5000079775
Encompass
MSD s U-PLEX GM-CSF Singleplex Assay is designed for non-human primate samples utilizing electrochemiluminescence detection technology for precise granulocyte-macrophage colony-stimulating factor measurement This research-grade immunoassay offers high sensitivity and specificity for cytokine analysis in preclinical studies supporting pharmaceutical and biomedical research applications requiring accurate GM-CSF quantification in primate biological samples
Catalog No. 50-000-79775 Supplier Meso Scale Discovery Supplier No. K156UMK21
MSD s U-PLEX GM-CSF Singleplex Assay is designed for non-human primate samples utilizing electrochemiluminescence detection technology for precise granulocyte-macrophage colony-stimulating factor measurement This research-grade immunoassay offers high sensitivity and specificity for cytokine analysis in preclinical studies supporting pharmaceutical and biomedical research applications requiring accurate GM-CSF quantification in primate biological samples