Learn More
Meso Scale Discovery U-PLEX NHP GRO- ASY 25 PL
SDP
Catalog No. 5000080075
Encompass
The Meso Scale Discovery U-PLEX NHP GRO alpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of GRO alpha in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring GRO alpha concentrations between 0 61-2500 pg/mL This assay provides a rapid reproducible method for quantifying GRO alpha levels with low sample volume requirements
Catalog No. 50-000-80075 Supplier Meso Scale Discovery Supplier No. K156UXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP GRO alpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of GRO alpha in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring GRO alpha concentrations between 0 61-2500 pg/mL This assay provides a rapid reproducible method for quantifying GRO alpha levels with low sample volume requirements