Learn More
Meso Scale Discovery U-PLEX NHP GRO- ASY
SDP
Catalog No. 5000080069
Encompass
The Meso Scale Discovery U-PLEX NHP GRO alpha Assay is a multiplex immunoassay kit for quantifying GRO alpha in human serum plasma and cell culture supernatants It uses MSD s SECTOR Imager 6000 for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for a standard curve and sample measurement It offers a streamlined workflow and results in 3 5 hours requiring minimal sample volume making it ideal for biomarker research and drug discovery
Catalog No. 50-000-80069 Supplier Meso Scale Discovery Supplier No. K156UXK1
The Meso Scale Discovery U-PLEX NHP GRO alpha Assay is a multiplex immunoassay kit for quantifying GRO alpha in human serum plasma and cell culture supernatants It uses MSD s SECTOR Imager 6000 for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for a standard curve and sample measurement It offers a streamlined workflow and results in 3 5 hours requiring minimal sample volume making it ideal for biomarker research and drug discovery