Learn More
Meso Scale Discovery U-PLEX NHP GRO- ASY384-well
SDP
Catalog No. 5000081751
Encompass
The Meso Scale Discovery U-PLEX NHP GRO alpha 384-well assay is a multiplex immunoassay kit that enables simultaneous quantification of human GRO alpha in cell culture supernatants serum plasma (EDTA heparin) and urine It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity and wide dynamic range Each kit provides reagents for one 384-well plate including antibody-coated plates calibrator and detection antibody Sample dilution is required prior to running the assay
Catalog No. 50-000-81751 Supplier Meso Scale Discovery Supplier No. K256UXK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP GRO alpha 384-well assay is a multiplex immunoassay kit that enables simultaneous quantification of human GRO alpha in cell culture supernatants serum plasma (EDTA heparin) and urine It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity and wide dynamic range Each kit provides reagents for one 384-well plate including antibody-coated plates calibrator and detection antibody Sample dilution is required prior to running the assay