Learn More
Meso Scale Discovery U-PLEX NHP GRO ASYQP
SDP
Catalog No. 5000080070
Encompass
The U-PLEX NHP GRO Assay (K156UXK) is a specialized immunoassay designed for quantitative measurement of Growth-Related Oncogene Alpha (GRO) in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for research applications studying inflammatory responses and chemokine signaling pathways in preclinical primate models
Catalog No. 50-000-80070 Supplier Meso Scale Discovery Supplier No. K156UXK21
The U-PLEX NHP GRO Assay (K156UXK) is a specialized immunoassay designed for quantitative measurement of Growth-Related Oncogene Alpha (GRO) in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for research applications studying inflammatory responses and chemokine signaling pathways in preclinical primate models