Learn More
Meso Scale Discovery U-PLEX NHP I-309 ASY 25 PL
SDP
Catalog No. 5000080027
Encompass
The U-PLEX NHP-I 309 Assay from Meso Scale Diagnostics is a multiplex immunoassay measuring nine biomarkers related to neuroinflammation and neurodegeneration It uses electrochemiluminescence detection on the QuickPlex SQ 120 instrument The analytes include A 1-42 A 1-40 -synuclein t-tau p-tau NFL GFAP sTREM2 and YKL-40 Requiring only 50 L of sample it offers high sensitivity and a broad dynamic range The kit includes a 96-well plate calibrators controls sample diluent detection antibodies read buffer and plate seals enabling efficient profiling of neurodegenerative disease biomarkers
Catalog No. 50-000-80027 Supplier Meso Scale Discovery Supplier No. K156UYK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP-I 309 Assay from Meso Scale Diagnostics is a multiplex immunoassay measuring nine biomarkers related to neuroinflammation and neurodegeneration It uses electrochemiluminescence detection on the QuickPlex SQ 120 instrument The analytes include A 1-42 A 1-40 -synuclein t-tau p-tau NFL GFAP sTREM2 and YKL-40 Requiring only 50 L of sample it offers high sensitivity and a broad dynamic range The kit includes a 96-well plate calibrators controls sample diluent detection antibodies read buffer and plate seals enabling efficient profiling of neurodegenerative disease biomarkers