Learn More
Meso Scale Discovery U-PLEX NHP I-309 ASY
SDP
Catalog No. 5000080035
Encompass
The Meso Scale Discovery U-PLEX NHP-I 309 Assay is a multiplex immunoassay panel measuring nine neurodegenerative and inflammatory biomarkers from serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection on Meso Scale Discovery s multi-array plate platform Researchers can measure A 42 A 40 t-tau p-tau -synuclein NFL GFAP sTREM2 and YKL-40 simultaneously from a small sample Its high sensitivity broad dynamic range and small sample needs make it ideal for clinical research and drug development in Alzheimer s Parkinson s ALS and other neurodegenerative diseases
Catalog No. 50-000-80035 Supplier Meso Scale Discovery Supplier No. K156UYK1
The Meso Scale Discovery U-PLEX NHP-I 309 Assay is a multiplex immunoassay panel measuring nine neurodegenerative and inflammatory biomarkers from serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection on Meso Scale Discovery s multi-array plate platform Researchers can measure A 42 A 40 t-tau p-tau -synuclein NFL GFAP sTREM2 and YKL-40 simultaneously from a small sample Its high sensitivity broad dynamic range and small sample needs make it ideal for clinical research and drug development in Alzheimer s Parkinson s ALS and other neurodegenerative diseases