Learn More
Meso Scale Discovery U-PLEX NHP I-309 ASY384-well
SDP
Catalog No. 5000078397
Encompass
The U-PLEX NHP-I 309-384 Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay panel that allows simultaneous measurement of up to 76 biomarkers related to neuroinflammation and neurodegeneration It utilizes electrochemiluminescent detection for high sensitivity and a broad dynamic range This assay panel includes cytokines chemokines growth factors and other soluble targets relevant to neurological diseases and disorders It requires only 25 L of sample per well and the kit provides reagents for testing 309 samples in 384-well plates
Catalog No. 50-000-78397 Supplier Meso Scale Discovery Supplier No. K256UYK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP-I 309-384 Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay panel that allows simultaneous measurement of up to 76 biomarkers related to neuroinflammation and neurodegeneration It utilizes electrochemiluminescent detection for high sensitivity and a broad dynamic range This assay panel includes cytokines chemokines growth factors and other soluble targets relevant to neurological diseases and disorders It requires only 25 L of sample per well and the kit provides reagents for testing 309 samples in 384-well plates