Learn More
Meso Scale Discovery U-PLEX NHP I-TAC ASY 25 PL
SDP
Catalog No. 5000080293
Encompass
The U-PLEX NHP-I TAC Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that measures total antioxidant capacity in human serum plasma and urine samples The kit contains a 96-well plate coated with a proprietary oxidant that reacts with antioxidants in the sample It utilizes MSD s patented SULFO-TAG labels that emit light when electrochemically stimulated The assay has a broad dynamic range from 1-2500 M Trolox equivalents with high sensitivity and reproducibility It requires a small sample volume and provides rapid results in under 4 hours The kit includes all necessary reagents and can process up to 80 samples in duplicate
Catalog No. 50-000-80293 Supplier Meso Scale Discovery Supplier No. K156UWK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP-I TAC Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that measures total antioxidant capacity in human serum plasma and urine samples The kit contains a 96-well plate coated with a proprietary oxidant that reacts with antioxidants in the sample It utilizes MSD s patented SULFO-TAG labels that emit light when electrochemically stimulated The assay has a broad dynamic range from 1-2500 M Trolox equivalents with high sensitivity and reproducibility It requires a small sample volume and provides rapid results in under 4 hours The kit includes all necessary reagents and can process up to 80 samples in duplicate