Learn More
Meso Scale Discovery U-PLEX NHP I-TAC ASY384-well
SDP
Catalog No. 5000078420
Encompass
The U-PLEX NHP-I TAC Assay by Meso Scale Diagnostics is a multiplex immunoassay that measures 10 neurodegenerative and inflammatory biomarkers from human serum plasma and CSF Using MSD s multi-array technology it provides sensitive reproducible results from small samples The panel includes total tau phosphorylated tau A 42 A 40 -synuclein neurogranin YKL-40 sTREM2 GFAP and NFL The assay kit offers an efficient workflow for profiling neurodegeneration and neuroinflammation biomarkers with a testing time of 2 5 hours for up to 80 samples per plate
Catalog No. 50-000-78420 Supplier Meso Scale Discovery Supplier No. K256UWK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP-I TAC Assay by Meso Scale Diagnostics is a multiplex immunoassay that measures 10 neurodegenerative and inflammatory biomarkers from human serum plasma and CSF Using MSD s multi-array technology it provides sensitive reproducible results from small samples The panel includes total tau phosphorylated tau A 42 A 40 -synuclein neurogranin YKL-40 sTREM2 GFAP and NFL The assay kit offers an efficient workflow for profiling neurodegeneration and neuroinflammation biomarkers with a testing time of 2 5 hours for up to 80 samples per plate