Learn More
Meso Scale Discovery U-PLEX NHP I-TAC ASYQP 25 PL
SDP
Catalog No. 5000080032
Encompass
The MSD U-PLEX I-TAC Singleplex Assay is a specialized immunoassay designed for measuring I-TAC (Interferon-inducible T-cell Alpha Chemoattractant) levels in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection The singleplex format allows focused analysis of I-TAC a chemokine involved in immune responses and inflammation pathways making it valuable for research applications in immunology and drug development studies using primate models
Catalog No. 50-000-80032 Supplier Meso Scale Discovery Supplier No. K156UWK24
Add to Cart
Add to Cart
Request a Quote
The MSD U-PLEX I-TAC Singleplex Assay is a specialized immunoassay designed for measuring I-TAC (Interferon-inducible T-cell Alpha Chemoattractant) levels in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection The singleplex format allows focused analysis of I-TAC a chemokine involved in immune responses and inflammation pathways making it valuable for research applications in immunology and drug development studies using primate models