Learn More
Meso Scale Discovery U-PLEX NHP IFN- 2a ASY  DFS Item
Catalog No. 5000080105
The Meso Scale Discovery U-PLEX NHP IFN alpha2a Assay Kit is a multiplex immunoassay for measuring human interferon alpha2a (IFN 2a) in various biological samples It uses MSD s electrochemiluminescent detection technology for quantitative measurement with high sensitivity and a wide dynamic range The assay requires 50 L of sample and is compatible with MSD instruments The kit includes a plate with capture antibodies detection antibodies calibrators and controls for accurate and precise IFN 2a quantification
Catalog No. 50-000-80105 Supplier Meso Scale Discovery Supplier No. K156VHK2
The Meso Scale Discovery U-PLEX NHP IFN alpha2a Assay Kit is a multiplex immunoassay for measuring human interferon alpha2a (IFN 2a) in various biological samples It uses MSD s electrochemiluminescent detection technology for quantitative measurement with high sensitivity and a wide dynamic range The assay requires 50 L of sample and is compatible with MSD instruments The kit includes a plate with capture antibodies detection antibodies calibrators and controls for accurate and precise IFN 2a quantification