Learn More
Meso Scale Discovery U-PLEX NHP IFN- 2a ASY
SDP
Catalog No. 5000080297
Encompass
The Meso Scale Discovery U-PLEX NHP IFN alpha2a Assay Kit is a multiplex immunoassay for measuring human interferon alpha 2a (IFN 2a) in various biological matrices It uses MSD s electrochemiluminescent detection for quantitative measurement with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and necessary calibrators and controls This assay offers a rapid reproducible method to quantify IFN 2a levels accurately and precisely
Catalog No. 50-000-80297 Supplier Meso Scale Discovery Supplier No. K156VHK1
The Meso Scale Discovery U-PLEX NHP IFN alpha2a Assay Kit is a multiplex immunoassay for measuring human interferon alpha 2a (IFN 2a) in various biological matrices It uses MSD s electrochemiluminescent detection for quantitative measurement with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and necessary calibrators and controls This assay offers a rapid reproducible method to quantify IFN 2a levels accurately and precisely