Learn More
Meso Scale Discovery U-PLEX NHP IFN- ASY 25 PL
SDP
Catalog No. 5000081393
Encompass
The Meso Scale Discovery U-PLEX NHP IFN gamma Assay is a multiplex immunoassay kit for measuring IFN gamma in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions It offers high sensitivity a broad dynamic range and requires only 25 L per well quantifying IFN gamma from 15 to 10 000 pg/mL
Catalog No. 50-000-81393 Supplier Meso Scale Discovery Supplier No. K156TTK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IFN gamma Assay is a multiplex immunoassay kit for measuring IFN gamma in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions It offers high sensitivity a broad dynamic range and requires only 25 L per well quantifying IFN gamma from 15 to 10 000 pg/mL