Learn More
Meso Scale Discovery U-PLEX NHP IFN- ASY
SDP
Catalog No. 5000079934
Encompass
The Meso Scale Discovery U-PLEX NHP IFN gamma Assay is an electrochemiluminescent immunoassay for measuring IFN gamma in non-human primate serum plasma and cell culture supernatant It uses MSD s SECTOR Imager offering high sensitivity broad dynamic range and low sample volume Detecting concentrations from 0 05-2500 pg/mL it has high specificity and minimal cross-reactivity The kit includes a 96-well plate antibodies buffers diluent and calibrators completing in about 3 5 hours It provides a sensitive specific high-throughput method for quantifying IFN gamma
Catalog No. 50-000-79934 Supplier Meso Scale Discovery Supplier No. K156TTK2
The Meso Scale Discovery U-PLEX NHP IFN gamma Assay is an electrochemiluminescent immunoassay for measuring IFN gamma in non-human primate serum plasma and cell culture supernatant It uses MSD s SECTOR Imager offering high sensitivity broad dynamic range and low sample volume Detecting concentrations from 0 05-2500 pg/mL it has high specificity and minimal cross-reactivity The kit includes a 96-well plate antibodies buffers diluent and calibrators completing in about 3 5 hours It provides a sensitive specific high-throughput method for quantifying IFN gamma