Learn More
Meso Scale Discovery U-PLEX NHP IFN- ASY384-well
SDP
Catalog No. 5000079465
Encompass
The Meso Scale Discovery U-PLEX NHP IFN gamma 384-well assay is a multiplex immunoassay panel that enables simultaneous quantification of IFN gamma in non-human primate samples such as cynomolgus rhesus and marmoset This 384-well plate assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It includes all necessary reagents and can process up to 40 samples in duplicate The panel is rigorously tested to ensure optimal performance and reliability With a streamlined and rapid workflow this assay allows efficient and accurate analysis of NHP IFN gamma
Catalog No. 50-000-79465 Supplier Meso Scale Discovery Supplier No. K256TTK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IFN gamma 384-well assay is a multiplex immunoassay panel that enables simultaneous quantification of IFN gamma in non-human primate samples such as cynomolgus rhesus and marmoset This 384-well plate assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It includes all necessary reagents and can process up to 40 samples in duplicate The panel is rigorously tested to ensure optimal performance and reliability With a streamlined and rapid workflow this assay allows efficient and accurate analysis of NHP IFN gamma