Learn More
Meso Scale Discovery U-PLEX NHP IFN ASYQP
SDP
Catalog No. 5000081551
Encompass
The U-PLEX NHP IFN Assay (K156TTK) is a specialized immunoassay designed for measuring interferon-gamma levels in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for cytokine quantification in research applications The assay utilizes MSD s proprietary U-PLEX technology enabling precise measurement of IFN concentrations in serum plasma and cell culture supernatants from non-human primate species making it ideal for preclinical immunology and vaccine research studies
Catalog No. 50-000-81551 Supplier Meso Scale Discovery Supplier No. K156TTK21
The U-PLEX NHP IFN Assay (K156TTK) is a specialized immunoassay designed for measuring interferon-gamma levels in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for cytokine quantification in research applications The assay utilizes MSD s proprietary U-PLEX technology enabling precise measurement of IFN concentrations in serum plasma and cell culture supernatants from non-human primate species making it ideal for preclinical immunology and vaccine research studies