Learn More
Meso Scale Discovery U-PLEX NHP IFN2a ASYQP
SDP
Catalog No. 5000080298
Encompass
The U-PLEX NHP IFN-alpha2a Assay (K156VHK) is a specialized immunoassay kit designed for quantitative measurement of interferon alpha-2a in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples providing researchers with a reliable tool for cytokine analysis in preclinical studies and immunology research involving primate models
Catalog No. 50-000-80298 Supplier Meso Scale Discovery Supplier No. K156VHK21
The U-PLEX NHP IFN-alpha2a Assay (K156VHK) is a specialized immunoassay kit designed for quantitative measurement of interferon alpha-2a in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit is optimized for serum plasma and cell culture supernatant samples providing researchers with a reliable tool for cytokine analysis in preclinical studies and immunology research involving primate models