Learn More
Meso Scale Discovery U-PLEX NHP IL-1 ASY 25 PL
SDP
Catalog No. 5000079891
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is an ultra-sensitive immunoassay that enables the detection of natural human IL-1alpha in serum plasma and cell culture supernatants This multiplex assay utilizes MSD s proprietary electrochemiluminescent detection technology and is compatible with commonly used sample matrices The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantify IL-1alpha levels down to sub-pg/mL concentrations This assay provides high sensitivity a wide dynamic range and requires a low sample volume of 50 L
Catalog No. 50-000-79891 Supplier Meso Scale Discovery Supplier No. K156UNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is an ultra-sensitive immunoassay that enables the detection of natural human IL-1alpha in serum plasma and cell culture supernatants This multiplex assay utilizes MSD s proprietary electrochemiluminescent detection technology and is compatible with commonly used sample matrices The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantify IL-1alpha levels down to sub-pg/mL concentrations This assay provides high sensitivity a wide dynamic range and requires a low sample volume of 50 L