Learn More
Meso Scale Discovery U-PLEX NHP IL-1 ASY
SDP
Catalog No. 5000080117
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in serum plasma and cell culture supernatants The assay uses MSD s proprietary QuickPlex SQ 120 instrument and U-PLEX linker-coated plates and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a plate detection antibody read buffer calibrator diluent wash buffer sample diluent and a vial of lyophilized calibrator that must be reconstituted before use The assay has high sensitivity with a lower limit of detection of 0 07 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-80117 Supplier Meso Scale Discovery Supplier No. K156UNK1
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in serum plasma and cell culture supernatants The assay uses MSD s proprietary QuickPlex SQ 120 instrument and U-PLEX linker-coated plates and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a plate detection antibody read buffer calibrator diluent wash buffer sample diluent and a vial of lyophilized calibrator that must be reconstituted before use The assay has high sensitivity with a lower limit of detection of 0 07 pg/mL and an upper limit of 10 000 pg/mL