Learn More
Meso Scale Discovery U-PLEX NHP IL-1 ASY
SDP
Catalog No. 5000080239
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is a sensitive immunoassay for detecting and quantifying natural human IL-1alpha in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay provides a simple rapid and robust method for measuring IL-1alpha levels
Catalog No. 50-000-80239 Supplier Meso Scale Discovery Supplier No. K156UNK2
The Meso Scale Discovery U-PLEX NHP IL-1alpha Assay Kit is a sensitive immunoassay for detecting and quantifying natural human IL-1alpha in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay provides a simple rapid and robust method for measuring IL-1alpha levels