Learn More
Meso Scale Discovery U-PLEX NHP IL-1 ASY384-well
SDP
Catalog No. 5000079342
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1alpha 384-well Assay Kit is a multiplex immunoassay that measures human IL-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of IL-1alpha and other biomarkers with minimal sample volume requirements The kit has high lot-to-lot and run-to-run reproducibility
Catalog No. 50-000-79342 Supplier Meso Scale Discovery Supplier No. K256UNK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1alpha 384-well Assay Kit is a multiplex immunoassay that measures human IL-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of IL-1alpha and other biomarkers with minimal sample volume requirements The kit has high lot-to-lot and run-to-run reproducibility