Learn More
Meso Scale Discovery U-PLEX NHP IL-1 ASYQP
SDP
Catalog No. 5000080756
Encompass
The U-PLEX NHP IL-1 Assay (K156UNK) is a multiplex immunoassay kit designed for quantitative measurement of interleukin-1 alpha in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying inflammatory responses and immune system function in preclinical primate models
Catalog No. 50-000-80756 Supplier Meso Scale Discovery Supplier No. K156UNK22
The U-PLEX NHP IL-1 Assay (K156UNK) is a multiplex immunoassay kit designed for quantitative measurement of interleukin-1 alpha in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying inflammatory responses and immune system function in preclinical primate models