Learn More
Meso Scale Discovery U-PLEX NHP IL-10 ASY 25 PL
SDP
Catalog No. 5000081237
Encompass
The Meso Scale Discovery U-PLEX NHP IL-10 Assay is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager offering a sensitive accurate method with a detection limit of 0 04 pg/mL and quantification up to 2500 pg/mL The assay can be multiplexed with other biomarkers and takes about 3 5 hours It employs a plate with capture antibodies and ruthenium-labeled detection antibodies optimized for MSD s instruments and software
Catalog No. 50-000-81237 Supplier Meso Scale Discovery Supplier No. K156TZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-10 Assay is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager offering a sensitive accurate method with a detection limit of 0 04 pg/mL and quantification up to 2500 pg/mL The assay can be multiplexed with other biomarkers and takes about 3 5 hours It employs a plate with capture antibodies and ruthenium-labeled detection antibodies optimized for MSD s instruments and software